Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073855.6 | NP_506256.3 | 2 - 3 | 682 | 87 | |
| GenBank mRNA | AY314773.1 | - | 2 - 3 | 675 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fmi-1| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073855.6 | NP_506256.3 | 2 - 3 | 682 | 87 | |
| GenBank mRNA | AY314773.1 | - | 2 - 3 | 675 | 87 | |
Set Membership: |
|