Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073855.6 | NP_506256.3 | 4 - 5 | 876 | 110 | |
| GenBank mRNA | AY314773.1 | - | 4 - 5 | 869 | 110 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map