Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 4 - 5 | 1247 | 86 | |
| NM_073855.6 | NP_506256.3 | 8 - 9 | 2351 | 86 | ||
| GenBank mRNA | AY314773.1 | - | 8 - 9 | 2344 | 86 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map