Assay Details
Gene Symbol: sun-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073880.6 | NP_506281.1 | 1 - 2 | 399 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
