Assay Details
| Gene Symbol: C50B6.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073902.4 | NP_506303.1 | 6 - 7 | 1477 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C50B6.7| Gene Symbol: C50B6.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073902.4 | NP_506303.1 | 6 - 7 | 1477 | 84 | |
Set Membership: |
|