Assay Details
| Gene Symbol: lgc-48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073907.3 | NP_506308.3 | 1 - 2 | 144 | 79 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Detect Genomic DNA  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 lgc-48
lgc-48 Genomic Map
Genomic Map Back To Top
 Back To Top