Assay Details
| Gene Symbol: gcy-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073918.6 | NP_506319.1 | 3 - 4 | 229 | 110 | |
| GenBank mRNA | AY652943.1 | - | 3 - 4 | 229 | 110 | |
More Information
Set Membership: |
|

gcy-34
Genomic Map