Assay Details
| Gene Symbol: F53F1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073922.5 | NP_506323.1 | 4 - 5 | 580 | 123 | |
| Gene Symbol: F53F1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383440.1 | 8 - 9 | 1669 | 123 | ||
More Information
Set Membership: |
|

F53F1.3
Genomic Map