Assay Details
| Gene Symbol: mes-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073932.6 | NP_506333.1 | 9 - 10 | 2495 | 57 | |
| GenBank mRNA | AF233290.1 | - | 9 - 10 | 2517 | 57 | |
More Information
Set Membership: |
|

mes-4
Genomic Map