Assay Details
Gene Symbol: cav-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269594.2 | NP_001256523.1 | 9 - 10 | 947 | 94 | |
NM_001269595.3 | NP_001256524.1 | 7 - 8 | 666 | 94 | ||
NM_001269596.3 | NP_001256525.1 | 8 - 9 | 780 | 94 | ||
NM_001269597.3 | NP_001256526.1 | 3 - 4 | 303 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
