Assay Details
| Gene Symbol: fmo-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269603.2 | NP_001256532.1 | 3 - 4 | 457 | 140 | |
| NR_069899.1 | 3 - 4 | 454 | 140 | |||
| GenBank mRNA | AJ581300.1 | - | 3 - 4 | 479 | 140 | |
More Information
Set Membership: |
|

fmo-4
Genomic Map