Assay Details
| Gene Symbol: fmo-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269603.2 | NP_001256532.1 | 6 - 7 | 1511 | 116 | |
| NR_069899.1 | 6 - 7 | 1508 | 116 | |||
| GenBank mRNA | AJ581300.1 | - | 6 - 7 | 1533 | 116 | |
More Information
Set Membership: |
|

fmo-4
Genomic Map