Assay Details
Gene Symbol: dct-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028706.7 | NP_001023877.1 | 10 - 11 | 1453 | 77 | |
NM_001028708.6 | NP_001023879.1 | 10 - 11 | 1461 | 77 | ||
NM_001028709.5 | NP_001023880.1 | 10 - 11 | 1461 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
