Assay Details
| Gene Symbol: dct-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028706.7 | NP_001023877.1 | 7 - 8 | 887 | 121 | |
| NM_001028707.6 | NP_001023878.1 | 7 - 8 | 893 | 121 | ||
| NM_001028708.6 | NP_001023879.1 | 7 - 8 | 895 | 121 | ||
| NM_001028709.5 | NP_001023880.1 | 7 - 8 | 895 | 121 | ||
More Information
Set Membership: |
|

dct-17
Genomic Map