Assay Details
| Gene Symbol: gcy-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074051.6 | NP_506452.5 | 6 - 7 | 1295 | 109 | |
| GenBank mRNA | AY652942.1 | - | 6 - 7 | 1241 | 109 | |
More Information
Set Membership: |
|

gcy-32
Genomic Map