Assay Details
| Gene Symbol: hda-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074198.9 | NP_506599.1 | 3 - 4 | 1176 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hda-1| Gene Symbol: hda-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074198.9 | NP_506599.1 | 3 - 4 | 1176 | 66 | |
Set Membership: |
|