Assay Details
| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 3 - 4 | 287 | 85 | |
| NR_069988.1 | 2 - 3 | 103 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rfc-1| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 3 - 4 | 287 | 85 | |
| NR_069988.1 | 2 - 3 | 103 | 85 | |||
Set Membership: |
|