Assay Details
| Gene Symbol: dpf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269738.3 | NP_001256667.1 | 10 - 11 | 2043 | 133 | |
| NM_074449.6 | NP_506850.1 | 10 - 11 | 2268 | 133 | ||
| NM_074450.6 | NP_506851.1 | 11 - 12 | 2216 | 133 | ||
More Information
Set Membership: |
|

dpf-1
Genomic Map