Assay Details
| Gene Symbol: dpf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269738.3 | NP_001256667.1 | 4 - 5 | 410 | 85 | |
| NM_074449.6 | NP_506850.1 | 4 - 5 | 635 | 85 | ||
| NM_074450.6 | NP_506851.1 | 5 - 6 | 583 | 85 | ||
More Information
Set Membership: |
|

dpf-1
Genomic Map