Assay Details
| Gene Symbol: gcy-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074700.1 | NP_507101.1 | 12 - 13 | 2934 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-20| Gene Symbol: gcy-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074700.1 | NP_507101.1 | 12 - 13 | 2934 | 84 | |
Set Membership: |
|