Assay Details
Gene Symbol: syp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074744.7 | NP_507145.2 | 5 - 6 | 2668 | 60 | |
GenBank mRNA | AF515833.1 | - | 5 - 6 | 1179 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
