Assay Details
Gene Symbol: immt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074840.7 | NP_507241.2 | 5 - 6 | 1487 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
