Assay Details
| Gene Symbol: dpy-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029095.4 | NP_001024266.1 | 4 - 5 | 486 | 76 | |
| NR_002407.1 | 4 - 5 | 321 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-21| Gene Symbol: dpy-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029095.4 | NP_001024266.1 | 4 - 5 | 486 | 76 | |
| NR_002407.1 | 4 - 5 | 321 | 76 | |||
Set Membership: |
|