Assay Details
| Gene Symbol: gck-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136446.4 | NP_001129918.1 | 5 - 6 | 1262 | 78 | |
| NM_001269833.2 | NP_001256762.1 | 6 - 7 | 2915 | 78 | ||
| NM_075116.6 | NP_507517.2 | 7 - 8 | 1733 | 78 | ||
| GenBank mRNA | AY741200.1 | - | 7 - 8 | 1839 | 78 | |
More Information
Set Membership: |
|

gck-3
Genomic Map