Assay Details
| Gene Symbol: fbxb-119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075185.5 | NP_507586.2 | 2 - 3 | 502 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fbxb-119| Gene Symbol: fbxb-119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075185.5 | NP_507586.2 | 2 - 3 | 502 | 128 | |
Set Membership: |
|