Assay Details
| Gene Symbol: C08E8.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075190.4 | NP_507591.2 | 2 - 3 | 129 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C08E8.1| Gene Symbol: C08E8.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075190.4 | NP_507591.2 | 2 - 3 | 129 | 98 | |
Set Membership: |
|