Assay Details
| Gene Symbol: dmd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269953.2 | NP_001256882.1 | 4 - 5 | 577 | 67 | |
| NM_001269954.4 | NP_001256883.1 | 2 - 3 | 341 | 67 | ||
More Information
Set Membership: |
|

dmd-3
Genomic Map