Assay Details
Gene Symbol: mlt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075511.6 | NP_507912.2 | 4 - 5 | 484 | 111 | |
GenBank mRNA | DQ645956.1 | - | 4 - 5 | 477 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
