Assay Details
Gene Symbol: F46B3.16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075584.2 | NP_507985.2 | 1 - 2 | 77 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F46B3.16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075584.2 | NP_507985.2 | 1 - 2 | 77 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |