Assay Details
Gene Symbol: mlt-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270009.3 | NP_001256938.1 | 11 - 12 | 6406 | 87 | |
NM_001270010.3 | NP_001256939.1 | 10 - 11 | 4828 | 87 | ||
NM_001270011.2 | NP_001256940.1 | 10 - 11 | 3567 | 87 | ||
NM_001270012.3 | NP_001256941.1 | 11 - 12 | 6379 | 87 | ||
NM_001270013.3 | NP_001256942.1 | 10 - 11 | 6044 | 87 | ||
NM_001270014.3 | NP_001256943.1 | 11 - 12 | 4600 | 87 | ||
NM_001270015.4 | NP_001256944.1 | 9 - 10 | 4194 | 87 | ||
NM_001270016.3 | NP_001256945.1 | 9 - 10 | 4138 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
