Assay Details
Gene Symbol: mlt-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270009.3 | NP_001256938.1 | 13 - 14 | 8393 | 103 | |
NM_001270010.3 | NP_001256939.1 | 12 - 13 | 6815 | 103 | ||
NM_001270011.2 | NP_001256940.1 | 12 - 13 | 5554 | 103 | ||
NM_001270012.3 | NP_001256941.1 | 13 - 14 | 8366 | 103 | ||
NM_001270013.3 | NP_001256942.1 | 12 - 13 | 8031 | 103 | ||
NM_001270014.3 | NP_001256943.1 | 13 - 14 | 6587 | 103 | ||
NM_001270015.4 | NP_001256944.1 | 11 - 12 | 6181 | 103 | ||
NM_001270016.3 | NP_001256945.1 | 11 - 12 | 6125 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
