Assay Details
Gene Symbol: mlt-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270009.3 | NP_001256938.1 | 8 - 9 | 3152 | 78 | |
NM_001270010.3 | NP_001256939.1 | 8 - 9 | 3152 | 78 | ||
NM_001270011.2 | NP_001256940.1 | 8 - 9 | 3175 | 78 | ||
NM_001270012.3 | NP_001256941.1 | 8 - 9 | 3125 | 78 | ||
NM_001270013.3 | NP_001256942.1 | 7 - 8 | 2790 | 78 | ||
NM_001270014.3 | NP_001256943.1 | 7 - 8 | 2489 | 78 | ||
NM_001270015.4 | NP_001256944.1 | 7 - 8 | 2518 | 78 | ||
NM_001270016.3 | NP_001256945.1 | 7 - 8 | 2462 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
