Assay Details
| Gene Symbol: elp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270021.4 | NP_001256950.1 | 10 - 11 | 1518 | 155 | |
| NM_001270022.3 | NP_001256951.1 | 10 - 11 | 1480 | 155 | ||
More Information
Set Membership: |
|

elp-1
Genomic Map