Assay Details
| Gene Symbol: elp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270021.4 | NP_001256950.1 | 9 - 10 | 1379 | 112 | |
| NM_001270022.3 | NP_001256951.1 | 9 - 10 | 1341 | 112 | ||
More Information
Set Membership: |
|

elp-1
Genomic Map