Assay Details
Gene Symbol: grl-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270026.2 | NP_001256955.1 | 2 - 3 | 615 | 123 | |
NM_001270027.3 | NP_001256956.1 | 2 - 3 | 493 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
