Assay Details
Gene Symbol: mrp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029548.6 | NP_001024719.2 | 12 - 13 | 3318 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
