Assay Details
Gene Symbol: mrp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029548.6 | NP_001024719.2 | 19 - 20 | 4546 | 91 | |
NM_001038464.4 | NP_001033553.1 | 19 - 20 | 4552 | 91 | ||
NM_075721.9 | NP_508122.2 | 20 - 21 | 4811 | 91 | ||
NM_171938.8 | NP_741702.1 | 19 - 20 | 4542 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
