Assay Details
Gene Symbol: tba-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076154.6 | NP_508555.2 | 2 - 3 | 309 | 92 |
Gene Symbol: lbp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383542.1 | 2 - 3 | 220 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383542.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
