Assay Details
| Gene Symbol: sul-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076159.4 | NP_508560.1 | 10 - 11 | 1775 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sul-1| Gene Symbol: sul-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076159.4 | NP_508560.1 | 10 - 11 | 1775 | 108 | |
Set Membership: |
|