Assay Details
| Gene Symbol: rgs-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029543.5 | NP_001024714.1 | 7 - 8 | 1407 | 68 | |
| NM_001029544.6 | NP_001024715.1 | 3 - 4 | 298 | 68 | ||
| NM_001373260.4 | NP_001360776.1 | 9 | 1581 | 68 | ||
| NM_001373261.4 | NP_001360777.1 | 8 - 9 | 1572 | 68 | ||
| NM_001373262.3 | NP_001360778.1 | 8 | 1492 | 68 | ||
| NM_076209.7 | NP_508610.3 | 8 - 9 | 1572 | 68 | ||
| GenBank mRNA | AB080201.1 | - | 7 - 8 | 1438 | 68 | |
| AY569308.1 | - | 8 - 9 | 1483 | 68 | ||
More Information
Set Membership: |
|

rgs-7
Genomic Map