Assay Details
| Gene Symbol: aex-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076240.8 | NP_508641.2 | 7 - 8 | 751 | 98 | |
| GenBank mRNA | FJ165552.1 | - | 7 - 8 | 641 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aex-4| Gene Symbol: aex-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076240.8 | NP_508641.2 | 7 - 8 | 751 | 98 | |
| GenBank mRNA | FJ165552.1 | - | 7 - 8 | 641 | 98 | |
Set Membership: |
|