Assay Details
| Gene Symbol: lin-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076283.6 | NP_508684.4 | 5 - 6 | 643 | 129 | |
| GenBank mRNA | AF133217.1 | - | 5 - 6 | 698 | 129 | |
More Information
Set Membership: |
|

lin-18
Genomic Map