Assay Details
Gene Symbol: mrp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375011.1 | NP_001362008.1 | 6 - 7 | 1159 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
