Assay Details
Gene Symbol: mltn-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076364.4 | NP_508765.2 | 6 - 7 | 1247 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: mltn-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076364.4 | NP_508765.2 | 6 - 7 | 1247 | 149 |
Set Membership: |
![]() ![]() ![]() ![]() |