Assay Details
| Gene Symbol: ham-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029192.6 | NP_001024363.2 | 2 - 3 | 344 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ham-2| Gene Symbol: ham-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029192.6 | NP_001024363.2 | 2 - 3 | 344 | 92 | |
Set Membership: |
|