Assay Details
| Gene Symbol: lon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076411.7 | NP_508812.3 | 9 - 10 | 1232 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lon-2| Gene Symbol: lon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076411.7 | NP_508812.3 | 9 - 10 | 1232 | 149 | |
Set Membership: |
|