Assay Details
Gene Symbol: mtm-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076487.4 | NP_508888.2 | 1 - 2 | 102 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
