Assay Details
| Gene Symbol: scav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076518.6 | NP_508919.2 | 2 - 3 | 318 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scav-1| Gene Symbol: scav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076518.6 | NP_508919.2 | 2 - 3 | 318 | 100 | |
Set Membership: |
|