Assay Details
| Gene Symbol: pks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330898.4 | NP_001317900.1 | 12 - 13 | 5232 | 100 | |
| NM_076522.3 | NP_508923.2 | 12 - 13 | 5232 | 100 | ||
More Information
Set Membership: |
|

pks-1
Genomic Map