Assay Details
| Gene Symbol: pks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330898.4 | NP_001317900.1 | 28 - 29 | 13466 | 94 | |
| NM_076522.3 | NP_508923.2 | 27 - 28 | 22970 | 94 | ||
More Information
Set Membership: |
|

pks-1
Genomic Map